Anti-PARK2 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-1865R
BOSSBS-1865REA 8800 NOK
BOSSBS-1865R
Anti-PARK2 Rabbit Polyclonal Antibody
Antistoffer
Functions within a multiprotein E3 ubiquitin ligase complex, catalyzing the covalent attachment of ubiquitin moieties onto substrate proteins, such as BCL2, SYT11, CCNE1, GPR37, RHOT1/MIRO1, MFN1, MFN2, STUB1, a 22 kDa O-linked glycosylated isoform of SNCAIP, SEPT5, TOMM2, USP3, ZNF746 and AIMP2. Mediates monoubiquitination as well as 'Lys-48'-linked and 'Lys-63'-linked polyubiquitination of substrates depending on the context. Participates in the removal and/or detoxification of abnormally folded or damaged protein by mediating 'Lys-63'-linked polyubiquitination of misfolded proteins suchMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol PARK2
Antigen-navn Parkin
Antigen synonymer Parkinson disease protein 2|PARK2|PRKN|Parkinson juvenile disease protein 2|Ubiquitin E3 ligase PRKN
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 5071
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Immunogen 390-440/465
Skjema liquid
Rensing Purified by Protein A.
Konsentrasjon 1 ug/ul
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR