Anti-STEAP1 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-1901R
BOSSBS-1901REA 8500 NOK
BOSSBS-1901R
Anti-STEAP1 Rabbit Polyclonal Antibody
Antistoffer
Metalloreductase that has the ability to reduce both Fe(3+) to Fe(2+) and Cu(2+) to Cu(1+). Uses NAD(+) as acceptor (By similarity).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: STEAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR