Anti-STEAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-1901R-A488
BOSSBS-1901R-A488EA 10450 NOK
BOSSBS-1901R-A488
Anti-STEAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Metalloreductase that has the ability to reduce both Fe(3+) to Fe(2+) and Cu(2+) to Cu(1+). Uses NAD(+) as acceptor (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STEAP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR