Anti-ABI1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-1961R-A680
BOSSBS-1961R-A680EA 10425 NOK
BOSSBS-1961R-A680
Anti-ABI1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
May act in negative regulation of cell growth and transformation by interacting with nonreceptor tyrosine kinases ABL1 and/or ABL2. May play a role in regulation of EGF-induced Erk pathway activation. Involved in cytoskeletal reorganization and EGFR signaling. Together with EPS8 participates in transduction of signals from Ras to Rac. In vitro, a trimeric complex of ABI1, EPS8 and SOS1 exhibits Rac specific guanine nucleotide exchange factor (GEF) activity and ABI1 seems to act as an adapter in the complex. Regulates ABL1/c-Abl-mediated phosphorylation of ENAH. Recruits WASF1 to lamellipodiaMore Product Information