Anti-ABI1 Rabbit Polyclonal Antibody (Cy5.5®)

Leverandør: Bioss
BS-1961R-CY5.5
BOSSBS-1961R-CY5.5EA 10550 NOK
BOSSBS-1961R-CY5.5
Anti-ABI1 Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
May act in negative regulation of cell growth and transformation by interacting with nonreceptor tyrosine kinases ABL1 and/or ABL2. May play a role in regulation of EGF-induced Erk pathway activation. Involved in cytoskeletal reorganization and EGFR signaling. Together with EPS8 participates in transduction of signals from Ras to Rac. In vitro, a trimeric complex of ABI1, EPS8 and SOS1 exhibits Rac specific guanine nucleotide exchange factor (GEF) activity and ABI1 seems to act as an adapter in the complex. Regulates ABL1/c-Abl-mediated phosphorylation of ENAH. Recruits WASF1 to lamellipodiaMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol ABI1
Antigen-navn Abl-Interactor 1
Environmentally Preferable
Antigen synonymer Spectrin SH3 domain-binding protein 1|Abl-binding protein 4|SSH3BP|Eps8 SH3 domain-binding protein|Abl interactor 1|Nap1-binding protein|E3B1|Abelson interactor 1|SSH3BP1|ABLBP4|NAP1BP|ABI-1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5.5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 10006
Isotype IgG
ImmunoKjemi Yes
Immunogen 140-190/508
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR