Anti-HMOX1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-2075R-A350
BOSSBS-2075R-A350EA 10550 NOK
BOSSBS-2075R-A350
Anti-HMOX1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
Heme oxygenase cleaves the heme ring at the alpha methene bridge to form biliverdin. Biliverdin is subsequently converted to bilirubin by biliverdin reductase. Under physiological conditions, the activity of heme oxygenase is highest in the spleen, where senescent erythrocytes are sequestrated and destroyed. Exhibits cytoprotective effects since excess of free heme sensitizes cells to undergo apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol HMOX1
Antigen-navn Heme Oxidase 1
Environmentally Preferable
Antigen synonymer HSP32|bK286B10|HO-1|HMOX1D
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 350
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 3162
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR