Anti-Heme Oxygenase 1, Heme Oxygenase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-2075R-A680
BOSSBS-2075R-A680EA 10525 NOK
BOSSBS-2075R-A680
Anti-Heme Oxygenase 1, Heme Oxygenase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Heme oxygenase cleaves the heme ring at the alpha methene bridge to form biliverdin. Biliverdin is subsequently converted to bilirubin by biliverdin reductase. Under physiological conditions, the activity of heme oxygenase is highest in the spleen, where senescent erythrocytes are sequestrated and destroyed. Exhibits cytoprotective effects since excess of free heme sensitizes cells to undergo apoptosis.

Type: Primary
Antigen: Heme Oxygenase 1, Heme Oxygenase 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/288
Host: Rabbit
Isotype: IgG
Reactivity:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR