Anti-Heme Oxygenase 1, Heme Oxygenase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-2075R-A750
BOSSBS-2075R-A750EA 10525 NOK
BOSSBS-2075R-A750
Anti-Heme Oxygenase 1, Heme Oxygenase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Heme oxygenase cleaves the heme ring at the alpha methene bridge to form biliverdin. Biliverdin is subsequently converted to bilirubin by biliverdin reductase. Under physiological conditions, the activity of heme oxygenase is highest in the spleen, where senescent erythrocytes are sequestrated and destroyed. Exhibits cytoprotective effects since excess of free heme sensitizes cells to undergo apoptosis.

Type: Primary
Antigen: Heme Oxygenase 1, Heme Oxygenase 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/288
Host: Rabbit
Isotype: IgG
Reactivity:More Product Information
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn Heme Oxygenase 1, Heme Oxygenase 2
Antigen synonymer bK286B1|HO|HSP32|HMOX1|HO-1|HO1|HMOX1D
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet Pig,Chicken,Human,Rat,Mouse
Vert Rabbit
Gen-ID 3162
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 101-200/288
Immunogen KLH conjugated synthetic peptide derived from human Heme Oxygenase 1
Skjema Liquid
Molekylvekt 32 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR