Anti-Integrin alpha 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-2095R-A680
BOSSBS-2095R-A680EA 10525 NOK
BOSSBS-2095R-A680
Anti-Integrin alpha 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Integrin alpha-1/beta-1 is a receptor for laminin and collagen. It recognises the proline-hydroxylated sequence G-F-P-G-E-R in collagen. Involved in anchorage-dependent, negative regulation of EGF-stimulated cell growth.

Type: Primary
Antigen: Integrin alpha 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1001-1179/1179
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn Integrin alpha 1
Antigen synonymer Integrin alpha-1|ITGA1|VLA-1|VLA1|Laminin and collagen receptor|CD49 antigen-like family member A|CD49a
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet Human,Rat
Vert Rabbit
Gen-ID 3672
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Modifikasjon Unmodified
Epitop 1001-1179/1179
Immunogen KLH conjugated synthetic peptide derived from human Integrin alpha 1
Skjema Liquid
Molekylvekt 128 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR