Anti-Peptide YY/PYY Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-2265R-CY7
BOSSBS-2265R-CY7EA 10550 NOK
BOSSBS-2265R-CY7
Anti-Peptide YY/PYY Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
This gut peptide inhibits exocrine pancreatic secretion, has a vasoconstrictory action and inhibitis jejunal and colonic mobility.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Peptide YY/PYY
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigen-navn Peptide YY/PYY
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy7®
Reaktivitet Human
Vert Rabbit
Gen-ID 5697
Isotype IgG
ImmunoKjemi Yes
Immunogen 20-70/97
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR