Anti-Peptide YY/PYY Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Bioss
BS-2265R-FITC
BOSSBS-2265R-FITCEA 10550 NOK
BOSSBS-2265R-FITC
Anti-Peptide YY/PYY Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
This gut peptide inhibits exocrine pancreatic secretion, has a vasoconstrictory action and inhibitis jejunal and colonic mobility.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Peptide YY/PYY
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR