Anti-GPD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-2388R-A750
BOSSBS-2388R-A750EA 10425 NOK
BOSSBS-2388R-A750
Anti-GPD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Glycerol 3 Phosphate Dehydrogenase is a homodimer and belongs to the NAD dependent Glycerol 3 Phosphate Dehydrogenase family.

Type: Primary
Antigen: GPD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/349
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR