Anti-GPD1 Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-2388R-CY5
BOSSBS-2388R-CY5EA 10550 NOK
BOSSBS-2388R-CY5
Anti-GPD1 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Glycerol 3 Phosphate Dehydrogenase is a homodimer and belongs to the NAD dependent Glycerol 3 Phosphate Dehydrogenase family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPD1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR