Anti-ACO2 Rabbit Polyclonal Antibody
Leverandør: Bioss
BS-2390R
BOSSBS-2390REA 8500 NOK
BOSSBS-2390R
Anti-ACO2 Rabbit Polyclonal Antibody
Antistoffer
ACO2, also referred to as aconitate hydratase, citrate hydrolyase or aconitase, is an iron-sulfur hydrolyase that catalyzes the non-limiting interconversion of citrate and isocitrate in the tricarboxylic acid cycle. It is expressed in the mitochondria and maintains a citrate:isocitrate ratio of approximately 10:1. ACO2 contains a redox-sensitive iron-sulfur cluster that exists in two states: active (Fe4S4) and inactive (Fe3S4). ACO2 activity is dependent on the state of this cluster as well as the presence of two conserved cysteine residues. In normal prostate epithelial cells ACO2 activityMore Product Information