Anti-ABP/SHBG Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-2410R-A750
BOSSBS-2410R-A750EA 10525 NOK
BOSSBS-2410R-A750
Anti-ABP/SHBG Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Functions as an androgen transport protein, but may also be involved in receptor mediated processes. Each dimer binds one molecule of steroid. Specific for 5-alpha-dihydrotestosterone, testosterone, and 17-beta-estradiol. Regulates the plasma metabolic clearance rate of steroid hormones by controlling their plasma concentration.

Type: Primary
Antigen: ABP/SHBG
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/402
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Bovine
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR