Anti-ATP2c1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-2434R-A350
BOSSBS-2434R-A350EA 10250 NOK
BOSSBS-2434R-A350
Anti-ATP2c1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of calcium.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATP2c1/2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR