Anti-ATP2c1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Leverandør: Bioss
BS-2434R-A555
BOSSBS-2434R-A555EA 10250 NOK
BOSSBS-2434R-A555
Anti-ATP2c1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antistoffer
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the transport of calcium.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATP2c1/2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol ATP2c1/2
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 555
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 27032, 9914
Isotype IgG
ImmunoKjemi Yes
Immunogen 315-352/919
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR