Anti-ATP2c1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-2434R-A680
BOSSBS-2434R-A680EA 10250 NOK
BOSSBS-2434R-A680
Anti-ATP2c1/2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
This magnesium-dependent enzyme catalyses the hydrolysis of ATP coupled with the transport of calcium.

Type: Primary
Antigen: ATP2c1/2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/919
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR