Anti-IL33 Rabbit Polyclonal Antibody (Cy5®)
Leverandør: Bioss
BS-2633R-CY5
BOSSBS-2633R-CY5EA 10650 NOK
BOSSBS-2633R-CY5
Anti-IL33 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Cytokine that binds to and signals through the IL1RL1/ST2 receptor which in turn activates NF-kappa-B and MAPK signaling pathways in target cells. Involved in the maturation of Th2 cells inducing the secretion of T-helper type 2-associated cytokines. Also involved in activation of mast cells, basophils, eosinophils and natural killer cells. Acts as a chemoattractant for Th2 cells, and may function as an "alarmin", that amplifies immune responses during tissue injury. In quiescent endothelia the uncleaved form is constitutively and abundantly expressed, and acts as a chromatin-associatedMore Product Information