Anti-IL33 Rabbit Polyclonal Antibody (Cy7®)
Leverandør: Bioss
BS-2633R-CY7
BOSSBS-2633R-CY7EA 10650 NOK
BOSSBS-2633R-CY7
Anti-IL33 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Cytokine that binds to and signals through the IL1RL1/ST2 receptor which in turn activates NF-kappa-B and MAPK signaling pathways in target cells. Involved in the maturation of Th2 cells inducing the secretion of T-helper type 2-associated cytokines. Also involved in activation of mast cells, basophils, eosinophils and natural killer cells. Acts as a chemoattractant for Th2 cells, and may function as an "alarmin", that amplifies immune responses during tissue injury. In quiescent endothelia the uncleaved form is constitutively and abundantly expressed, and acts as a chromatin-associatedMore Product Information
Bestill nå
SPESIFIKASJONER
Antigensymbol IL33
Environmentally Preferable
Antigen-navn Interleukin 33
Antigen synonymer NFEHEV|C9orf26|IL1F11|NF-HEV|Propeptide|DVS27
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy7®
Reaktivitet Rat,Mouse
Vert Rabbit
Gen-ID 77125
Isotype IgG
ImmunoKjemi Yes
Immunogen 214-264/266
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C