Anti-SPHK1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-2652R-A750
BOSSBS-2652R-A750EA 10525 NOK
BOSSBS-2652R-A750
Anti-SPHK1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Catalyses the phosphorylation of sphingosine to form sphingosine 1-phosphate (SPP), a lipid mediator with both intra- and extracellular functions. Also acts on D-erythro-sphingosine and to a lesser extent sphinganine, but not other lipids, such as D,L-threo-dihydrosphingosine, N,N-dimethylsphingosine, diacylglycerol, ceramide, or phosphatidylinositol.

Type: Primary
Antigen: SPHK1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/384
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestill nå

SPESIFIKASJONER

Antigen-navn SPHK1
Antigen synonymer SPHK|SK 1|SPHK1|SPK|Sphingosine kinase 1|SPK 1
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 8877
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Modifikasjon Unmodified
Epitop 1-100/384
Immunogen KLH conjugated synthetic peptide derived from human SPHK1
Skjema Liquid
Molekylvekt 42-52 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR