Anti-LTB4R2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-2655R-A350
BOSSBS-2655R-A350EA 10550 NOK
BOSSBS-2655R-A350
Anti-LTB4R2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
Low-affinity receptor for leukotrienes including leukotriene B4. Mediates chemotaxis of granulocytes and macrophages. The response is mediated via G-proteins that activate a phosphatidylinositol-calcium second messenger system. The rank order of affinities for the leukotrienes is LTB4 >12-epi-LTB4 >LTB5 >LTB3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LTB4R2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol LTB4R2
Environmentally Preferable
Antigen-navn Leukotriene B4 Receptor 2
Antigen synonymer NOP9|BLTR2|KPG_004|LTB4-R 2|JULF2|LTB4-R2|BLT2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 350
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 56413
Isotype IgG
ImmunoKjemi Yes
Immunogen 1-55/389
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR