Anti-LTB4R2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-2655R-A488
BOSSBS-2655R-A488EA 10550 NOK
BOSSBS-2655R-A488
Anti-LTB4R2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Low-affinity receptor for leukotrienes including leukotriene B4. Mediates chemotaxis of granulocytes and macrophages. The response is mediated via G-proteins that activate a phosphatidylinositol-calcium second messenger system. The rank order of affinities for the leukotrienes is LTB4 >12-epi-LTB4 >LTB5 >LTB3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LTB4R2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR