Anti-LTB4-R2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-2655R-A680
BOSSBS-2655R-A680EA 10525 NOK
BOSSBS-2655R-A680
Anti-LTB4-R2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Low-affinity receptor for leukotrienes including leukotriene B4. Mediates chemotaxis of granulocytes and macrophages. The response is mediated via G-proteins that activate a phosphatidylinositol-calcium second messenger system. The rank order of affinities for the leukotrienes is LTB4 > 12-epi-LTB4 > LTB5 > LTB3.

Type: Primary
Antigen: LTB4-R2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 25-120/389
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Bestill nå

SPESIFIKASJONER

Antigen-navn LTB4-R2
Environmentally Preferable
Antigen synonymer NOP9|Leukotriene B4 receptor BLT2|Seven transmembrane receptor BLTR2|BLTR2|KPG_004|LTB4 receptor JULF2|LTB4-R 2|JULF2|Leukotriene B4 receptor 2|LTB4-R2|BLT2R|LTB4R2|BLT2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet Rat,Mouse
Vert Rabbit
Gen-ID 56413
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Modifikasjon Unmodified
Epitop 25-120/389
Immunogen KLH conjugated synthetic peptide derived from human LTB4-R2
Skjema Liquid
Molekylvekt 41 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR