Anti-JAK3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverandør: Bioss
BS-2808R-A488
BOSSBS-2808R-A488EA 10550 NOK
BOSSBS-2808R-A488
Anti-JAK3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Non-receptor tyrosine kinase involved in various processes such as cell growth, development, or differentiation. Mediates essential signaling events in both innate and adaptive immunity and plays a crucial role in hematopoiesis during T-cells development. In the cytoplasm, plays a pivotal role in signal transduction via its association with type I receptors sharing the common subunit gamma such as IL2R, IL4R, IL7R, IL9R, IL15R and IL21R. Following ligand binding to cell surface receptors, phosphorylates specific tyrosine residues on the cytoplasmic tails of the receptor, creating dockingMore Product Information