Anti-Jak3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-2808R-A680
BOSSBS-2808R-A680EA 10525 NOK
BOSSBS-2808R-A680
Anti-Jak3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
Non-receptor tyrosine kinase involved in various processes such as cell growth, development, or differentiation. Mediates essential signaling events in both innate and adaptive immunity and plays a crucial role in hematopoiesis during T-cells development. In the cytoplasm, plays a pivotal role in signal transduction via its association with type I receptors sharing the common subunit gamma such as IL2R, IL4R, IL7R, IL9R, IL15R and IL21R. Following ligand binding to cell surface receptors, phosphorylates specific tyrosine residues on the cytoplasmic tails of the receptor, creating dockingMore Product Information