Anti-CRK Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Leverandør: Bioss
BS-3121R-A350
BOSSBS-3121R-A350EA 10800 NOK
BOSSBS-3121R-A350
Anti-CRK Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antistoffer
The Crk-I and Crk-II forms differ in their biological activities. Crk-II has less transforming activity than Crk-I. Crk-II mediates attachment-induced MAPK8 activation, membrane ruffling and cell motility in a Rac-dependent manner. Involved in phagocytosis of apoptotic cells and cell motility via its interaction with DOCK1 and DOCK4. May regulate the EFNA5-EPHA3 signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRK
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR