Anti-CRK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Bioss
BS-3121R-HRP
BOSSBS-3121R-HRPEA 10875 NOK
BOSSBS-3121R-HRP
Anti-CRK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
The Crk-I and Crk-II forms differ in their biological activities. Crk-II has less transforming activity than Crk-I. Crk-II mediates attachment-induced MAPK8 activation, membrane ruffling and cell motility in a Rac-dependent manner. Involved in phagocytosis of apoptotic cells and cell motility via its interaction with DOCK1 and DOCK4. May regulate the EFNA5-EPHA3 signaling.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CRK
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol CRK
Environmentally Preferable
Antigen-navn V-Crk Avian Sarcoma Virus Ct10 Oncogene Homolog
Antigen synonymer proto-oncogene c-Crk|CRKII|p38|adapter molecule crk
Antistofftype Primary
Klonalitet Polyclonal
Konjugering HRP (Horseradish Peroxidase)
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 1398
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Modifikasjon phospho-Tyr221
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR