Anti-GSK3B Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-3148R-CY7
BOSSBS-3148R-CY7EA 10600 NOK
BOSSBS-3148R-CY7
Anti-GSK3B Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Glycogen synthase kinase 3 (GSK3) is a proline directed serine threonine kinase that was initially identified as a phosphorylating and inactivating glycogen synthase. GSK3 has been implicated in fundamental cell processes such as cell fate determination, metabolism, transcriptional control and oncogenesis. Two isoforms, alpha (GSK3A; OMIM 606784) and beta, show a high degree of amino acid homology within their catalytic domains. GSK3B is involved in energy metabolism, neuronal cell development and body pattern formation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen:More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol GSK3B
Antigen-navn Glycogen Synthase Kinase 3 beta
Environmentally Preferable
Antigen synonymer EC 2.7.11.26|GSK3B|Factor A|GSK-3 beta Factor A|GSK-3 beta
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy7®
Reaktivitet Rabbit,Human
Vert Rabbit
Gen-ID 2932
Isotype IgG
ImmunoKjemi Yes
Modifikasjon phospho-Thr390
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR