Anti-GRIN2A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Leverandør: Bioss
BS-3305R-A647
BOSSBS-3305R-A647EA 10800 NOK
BOSSBS-3305R-A647
Anti-GRIN2A Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antistoffer
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate-gated ion channels. These receptors have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C) and NMDAR2D (GRIN2D). Alternatively spliced transcript variants encoding different isoforms have been found forMore Product Information