Anti-GRIN2A Rabbit Polyclonal Antibody (Cy5.5®)
Leverandør: Bioss
BS-3305R-CY5.5
BOSSBS-3305R-CY5.5EA 10875 NOK
BOSSBS-3305R-CY5.5
Anti-GRIN2A Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate-gated ion channels. These receptors have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C) and NMDAR2D (GRIN2D). Alternatively spliced transcript variants encoding different isoforms have been found forMore Product Information