Anti-NMDAR2B Tyr1070 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-3306R-A750
BOSSBS-3306R-A750EA 10750 NOK
BOSSBS-3306R-A750
Anti-NMDAR2B Tyr1070 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
NMDA receptor subtype of glutamate-gated ion channels with high calcium permeability and voltage-dependent sensitivity to magnesium. Mediated by glycine. In concert with DAPK1 at extrasynaptic sites, acts as a central mediator for stroke damage. Its phosphorylation at Ser-1303 by DAPK1 enhances synaptic NMDA receptor channel activity inducing injurious Ca2+ influx through them, resulting in an irreversible neuronal death (By similarity).

Type: Primary
Antigen: NMDAR2B Tyr1070
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn NMDAR2B Tyr1070
Antigen synonymer hNR3|N-methyl D-aspartate receptor subtype 2B|Glutamate [NMDA] receptor subunit epsilon-2|GRIN2B|N-methyl-D-aspartate receptor subunit 3|GluN2B|NR3|NR2B|MRD6|Glutamate receptor ionotropic, NMDA 2B|NMDAR2B
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 750
Reaktivitet
Vert Rabbit
Gen-ID 2904
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Phosphorylation (Tyr1070)
Immunogen KLH conjugated synthetic phosphopeptide derived from human NMDAR2B around the phosphorylation site of Tyr1070
Skjema Liquid
Molekylvekt 164 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR