Anti-NMDAR2B Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-3307R-A750
BOSSBS-3307R-A750EA 15800 NOK
BOSSBS-3307R-A750
Anti-NMDAR2B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
NMDA receptor subtype of glutamate-gated ion channels with high calcium permeability and voltage-dependent sensitivity to magnesium. Mediated by glycine. In concert with DAPK1 at extrasynaptic sites, acts as a central mediator for stroke damage. Its phosphorylation at Ser-133 by DAPK1 enhances synaptic NMDA receptor channel activity inducing injurious Ca2+ influx through them, resulting in an irreversible neuronal death (By similarity).

Type: Primary
Antigen: NMDAR2B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1431-1482/1482
Host: Rabbit
Isotype: IgG
More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR