Anti-RCC1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Leverandør: Bioss
BS-3360R-HRP
BOSSBS-3360R-HRPEA 10800 NOK
BOSSBS-3360R-HRP
Anti-RCC1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Ran GTPase plays important roles in nucleocytoplasmic transport in interphase and in both spindle formation and nuclear envelope (NE) assembly during mitosis. The latter functions rely on the presence of high local concentrations of GTP bound Ran near mitotic chromatin. RanGTP localization has been proposed to result from the association of Ran's GDP/GTP exchange factor, RCC1, with chromatin , but Ran is shown here to bind directly to chromatin in two modes, either dependent or independent of RCC1, and, where bound, to increase the affinity of chromatin for NE membranes.
RecommendedMore Product Information