Anti-SRGAP3 Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-3660R
BOSSBS-3660REA 8500 NOK
BOSSBS-3660R
Anti-SRGAP3 Rabbit Polyclonal Antibody
Antistoffer
srGAP3 (SLIT ROBO Rho GTPase-activating protein 3) is a GTPase activating protein (GAP) for the small G protein Rac. It has a putative role in attenuating Rac1 signalling in neurons. srGAP3 is thought to act downstream of the Slit-Robo pathway, which regulates neuronal migration and axonal branching, and play an important role in axonal regeneration after axotomy. It also plays a putative role in severe mental retardation.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SRGAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR