Anti-ALDH1A2 Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-3676R-CY7
BOSSBS-3676R-CY7EA 10350 NOK
BOSSBS-3676R-CY7
Anti-ALDH1A2 Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
Recognizes as substrates free retinal and cellular retinol-binding protein-bound retinal. Does metabolize octanal and decanal but does not metabolize citral, benzaldehyde, acetaldehyde and propanal efficiently (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALDH1A2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR