Anti-H3F3A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-3773R-A488
BOSSBS-3773R-A488EA 10800 NOK
BOSSBS-3773R-A488
Anti-H3F3A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Modulation of the chromatin structure plays an important role in the regulation of transcription in eukaryotes. The nucleosome, made up of four core histone proteins (H2A, H2B, H3 and H4), is the primary building block of chromatin. The N-terminal tail of core histones undergoes different posttranslational modifications including acetylation, phosphorylation and methylation. These modifications occur in response to cell signal stimuli and have a direct effect on gene expression. In most species, the histone H2B is primarily acetylated at lysines 5, 12, 15 and 20. Histone H3 is primarilyMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol H3F3A
Antigen-navn H3 histone, Family 3A
Environmentally Preferable
Antigen synonymer H3.3A|H3F3
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 488
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 3020
Isotype IgG
ImmunoKjemi Yes
Modifikasjon trimethyl-Lys27
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR