Anti-PNMT Rabbit Polyclonal Antibody (Cy3®)

Leverandør: Bioss
BS-3912R-CY3
BOSSBS-3912R-CY3EA 10350 NOK
BOSSBS-3912R-CY3
Anti-PNMT Rabbit Polyclonal Antibody (Cy3®)
Antistoffer
Converts noradrenaline to adrenaline.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PNMT
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR