Anti-PNMT Rabbit Polyclonal Antibody (Cy5®)

Leverandør: Bioss
BS-3912R-CY5
BOSSBS-3912R-CY5EA 10350 NOK
BOSSBS-3912R-CY5
Anti-PNMT Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Converts noradrenaline to adrenaline.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PNMT
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol PNMT
Antigen-navn Phenylethanolamine N-methyltransferase
Antigen synonymer PENT|PNMTase
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 5409
Isotype IgG
ImmunoKjemi Yes
Immunogen 130-180/282
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR