Anti-STARD5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Leverandør: Bioss
BS-3919R-A750
BOSSBS-3919R-A750EA 10425 NOK
BOSSBS-3919R-A750
Anti-STARD5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
STARD5, or START domain containing 5, belongs to a family containing a 200- to 210-amino acid STAR-related lipid transfer (START) domain. STARD5 may be involved in the intracellular transport of sterols or other lipids and may bind cholesterol or other sterols.

Type: Primary
Antigen: STARD5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/213
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Others
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR