Anti-STARD5 Rabbit Polyclonal Antibody (Cy5.5®)

Leverandør: Bioss
BS-3919R-CY5.5
BOSSBS-3919R-CY5.5EA 10550 NOK
BOSSBS-3919R-CY5.5
Anti-STARD5 Rabbit Polyclonal Antibody (Cy5.5®)
Antistoffer
STARD5, or START domain containing 5, belongs to a family containing a 200- to 210-amino acid STAR-related lipid transfer (START) domain. STARD5 may be involved in the intracellular transport of sterols or other lipids and may bind cholesterol or other sterols.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STARD5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol STARD5
Environmentally Preferable
Antigen-navn StAR-related lipid transfer (START) Domain Containing 5
Antigen synonymer StAR related lipid transfer protein 5|START domain containing 5|MGC10327.|StARD5|START domain containing protein 5
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy5.5®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 80765
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR