Anti-PKA gamma Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Leverandør: Bioss
BS-3964R-A680
BOSSBS-3964R-A680EA 10425 NOK
BOSSBS-3964R-A680
Anti-PKA gamma Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
PKA (or cAPK) is a cyclic AMP dependent protein kinase. When activated by the second messenger cAMP, PKA mediates diverse cellular mechanisms, including proliferation, ion transport, regulation of metabolism, plus gene transcription. PKA is comprised of two dimers of two subunits, R (regulatory) and C (catalytic). Two families of R subunit (RI and RII) and three C subunit isoforms (C alpha, C beta, and C gamma) have been identified each possessing distinct cAMP binding properties and resulting in different phosphorylation states. C subunit is activated through autophosphorylation and directMore Product Information