Anti-PRKACG Rabbit Polyclonal Antibody (Cy5®)
Leverandør: Bioss
BS-3964R-CY5
BOSSBS-3964R-CY5EA 10550 NOK
BOSSBS-3964R-CY5
Anti-PRKACG Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
PKA (or cAPK) is a cyclic AMP dependent protein kinase. When activated by the second messenger cAMP, PKA mediates diverse cellular mechanisms, including proliferation, ion transport, regulation of metabolism, plus gene transcription. PKA is comprised of two dimers of two subunits, R (regulatory) and C (catalytic). Two families of R subunit (RI and RII) and three C subunit isoforms (C alpha, C beta, and C gamma) have been identified each possessing distinct cAMP binding properties and resulting in different phosphorylation states. C subunit is activated through autophosphorylation and directMore Product Information