Anti-PRKACG Rabbit Polyclonal Antibody (Cy7®)

Leverandør: Bioss
BS-3964R-CY7
BOSSBS-3964R-CY7EA 10550 NOK
BOSSBS-3964R-CY7
Anti-PRKACG Rabbit Polyclonal Antibody (Cy7®)
Antistoffer
PKA (or cAPK) is a cyclic AMP dependent protein kinase. When activated by the second messenger cAMP, PKA mediates diverse cellular mechanisms, including proliferation, ion transport, regulation of metabolism, plus gene transcription. PKA is comprised of two dimers of two subunits, R (regulatory) and C (catalytic). Two families of R subunit (RI and RII) and three C subunit isoforms (C alpha, C beta, and C gamma) have been identified each possessing distinct cAMP binding properties and resulting in different phosphorylation states. C subunit is activated through autophosphorylation and directMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol PRKACG
Environmentally Preferable
Antigen-navn Protein Kinase cAMP-activated Catalytic Subunit Gamma
Antigen synonymer KAPG|PRKACG|PKA C gamma|KAPCG_HUMAN.|Protein kinase cAMP dependent catalytic gamma|Serine threonine protein kinase
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Cy7®
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Gen-ID 5568
Isotype IgG
ImmunoKjemi Yes
Skjema liquid
Rensing Purified by Protein A
Konsentrasjon 1 μg/μl
Kyssabsorpsjon No
Lagringsbuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagringstemp Store at 4°C for 12 months
Frakttemp. 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR