Anti-PAK6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Leverandør: Bioss
BS-4153R-A488
BOSSBS-4153R-A488EA 10450 NOK
BOSSBS-4153R-A488
Anti-PAK6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
This gene encodes a protein that shares a high degree of sequence similarity with p21-activated kinase (PAK) family members. The proteins of this family are Rac/Cdc42-associated Ste20-like Ser/Thr protein kinases, characterized by a highly conserved amino-terminal Cdc42/Rac interactive binding (CRIB) domain and a carboxyl-terminal kinase domain. PAK kinases are implicated in the regulation of a number of cellular processes, including cytoskeleton rearrangement, apoptosis and the MAP kinase signaling pathway. The protein encoded by this gene was found to interact with androgen receptor (AR),More Product Information