Anti-Sphingomyelin Synthase 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Leverandør: Bioss
BS-4216R-A750
BOSSBS-4216R-A750EA 10425 NOK
BOSSBS-4216R-A750
Anti-Sphingomyelin Synthase 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antistoffer
Sphingomyelin synthases synthesize the sphingolipid, sphingomyelin, through transfer of the phosphatidyl head group, phosphatidylcholine, on to the primary hydroxyl of ceramide. The reaction is bidirectional depending on the respective levels of the sphingolipid and ceramide. Golgi apparatus SMS1 directly and specifically Recognises the choline head group on the substrate, requiring two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. Major form in macrophages. Required for cell growth in certain cell types such as HeLa cells. Suppresses BAX-More Product Information