Anti-SGMS1 Rabbit Polyclonal Antibody (Cy5®)
Leverandør: Bioss
BS-4216R-CY5
BOSSBS-4216R-CY5EA 10550 NOK
BOSSBS-4216R-CY5
Anti-SGMS1 Rabbit Polyclonal Antibody (Cy5®)
Antistoffer
Sphingomyelin synthases synthesize the sphingolipid, sphingomyelin, through transfer of the phosphatidyl head group, phosphatidylcholine, on to the primary hydroxyl of ceramide. The reaction is bidirectional depending on the respective levels of the sphingolipid and ceramide. Golgi apparatus SMS1 directly and specifically recognizes the choline head group on the substrate, requiring two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. Major form in macrophages. Required for cell growth in certain cell types such as HeLa cells. Suppresses BAX-More Product Information