Anti-UGT2B4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Leverandør: Bioss
BS-4225R-A680
BOSSBS-4225R-A680EA 10425 NOK
BOSSBS-4225R-A680
Anti-UGT2B4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antistoffer
The UDP-Glucuronosyltransferases (UGT) comprise a family of enzymes that detoxify and enhance the urinary excretion of a wide variety of xenobiotic and endogenous substrates by transferring glucuronic acid to sulfhydryl, hydroxyl, aromatic amino, or carboxylic acid groups. They have been subdivided into two families, UGT1 and UGT2, based on the evolutionary divergence of their genes. The enzymes of the UGT1A family play an important role in the metabolism of dietary constituents, phenols, and therapeutic drugs, and also the glucuronidation of bilirubin and iodothyronines. The enzymes of theMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn UGT2B4
Environmentally Preferable
Antigen synonymer UDP glucuronosyltransferase 2B4 precursor|UD2B4_HUMAN|UGT2B11|UDP glycosyltransferase 2 family polypeptide B4|UDPGTh 1|UDP glucuronosyltransferase 2 family polypeptide B4|UDP glycosyltransferase|UDP glucuronyltransferase family 2 beta 4|UDPGTh1|Hyodeoxycholic acid|UDPGT|HLUG25|UGT2B4
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Alexa Fluor® 680
Reaktivitet
Vert Rabbit
Gen-ID 7363
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Modifikasjon Unmodified
Epitop 111-210/528
Immunogen KLH conjugated synthetic peptide derived from human UGT2B4
Skjema Liquid
Molekylvekt 58 kDa
Rensing Purified by Protein A
Konsentrasjon 1 µg/µl
Lagringsbuffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Lagringstemp Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Frakttemp. 4 °C
Uttestet applikasjon ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR