Anti-FMDV Polyprotein VP1 protein Rabbit Polyclonal Antibody

Leverandør: Bioss
BS-4521R
BOSSBS-4521REA 8500 NOK
BOSSBS-4521R
Anti-FMDV Polyprotein VP1 protein Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: FMDV Polyprotein VP1 protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR