Anti-CUEDC2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Leverandør: Bioss
BS-4608R-A488
BOSSBS-4608R-A488EA 10450 NOK
BOSSBS-4608R-A488
Anti-CUEDC2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antistoffer
Down-regulates ESR1 protein levels through the ubiquitination-proteasome pathway, regardless of the presence of 17 beta-estradiol. Also involved in 17 beta-estradiol-induced ESR1 degradation. Controls PGR protein levels through a similar mechanism.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CUEDC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR